missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Protein A - Sepharose™ 4B
Click to view available options
Quantity:
5 mL
25 mL
1 L
Unit Size:
1 L
25mL
5mL
Description
This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.
This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).
Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).
Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
Specifications
Specifications
| Product Type | Protein A Sepharose |
| Product Line | Sepharose |
| Column Type | Affinity |
| Stationary Phase | Protein A |
| Content And Storage | This product is supplied as a 50% suspension (25 ml Protein A-Sepharose™ 4B + 25 ml PBS) Wet Ice |
| Quantity | 25 mL |
| Form | Suspension |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction