missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Protein Expression Lysate Control Kit (Raji)

Product Code. 10239524 Shop All Applied Biosystems Products
Change view
Click to view available options
:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10239524 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10239524 Supplier Applied Biosystems™ Supplier No. 4405448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein Expression Lysate Control Kit (Raji)

The Applied Biosystems™ Raji Cell Lysate is a preparation from the Raji human B-cell line that has been characterized for use as an expression control lysate in TaqMan™ Protein Expression Assay experiments. The cell lysate was prepared as described in the Protein Expression Sample Preparation Kit Protocol (PN 4405785) using the Protein Expression Sample Preparation Kit (PN 4405443). The Raji Protein Expression Lysate Control Kit provides a non-stem cell line lysate that may be used as a positive control for ICAM1 and CTSB expression. The lysate contains protease and phosphatase inhibitors and has not been subjected to denaturing or reducing conditions.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes Lysate Control (store at<-50°C) and Lysate Dilution Buffer (store at -15 to -25°C).
Kit Type Protein Expression Lysate Control Kit
Product Line TaqMan
Shipping Condition Dry Ice
Species Human
For Use With (Application) Protein Expression Analysis, Cell Analysis
No. of Reactions 100 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.