missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein G HP SpinTrap™ Columns

Product Code. 11580644 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
800 μL
Unit Size:
100g
Product Code. Quantity unitSize
11580644 800 μL 100g
1 options
This item is not returnable. View return policy

Product Code. 11580644

Brand: Cytiva 28903134

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

16 prepacked Protein G HP SpinTrap columns, one plastic bottom cap removal tool, and instructions. Also included is a protocol, with no pretreatment of sample required, for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants in less than 20 min.

For small-scale purification of antibodies or enrichment of proteins of interest in prepacked spin columns

  • Protocols for both classic elution and cross-link methods; designed for use with a standard microcentrifuge
  • Formatted for elution for both electrophoresis and LC-MS analysis workflows
  • Fast-binding kinetics and high capacity provide high yield
  • Prepacked with Protein G Sepharose™ High Performance Medium for coupling of antibodies of IgG subclasses
  • Easy scale-up with HiTrap™ Protein G HP prepacked columns

  • Matrix: Highly cross-linked agarose, 6%
  • Binding capacity: >1mg human IgG/column
  • Average particle size: 34μm
  • pH stability: 3 to 9 (long term); 2 to 9 (short term)
  • Column volume: 800μL
  • Max. sample loading volume: 600μL

TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein G Sepharose HP SpinTrap spin column
Quantity 800 μL
Includes 16 Prepacked Protein G HP SpinTrap column, one Plastic botto m cap removal to ol and instructions, proto col
Ligand Type Recombinant Protein G lacking Albumin-binding region (2mg/mL medium)
pH Range 3 to 9 (long term), 2 to 9 (short term)
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 8°C
Column Type Protein G Sepharose Spin Column
For Use With (Application) Small-scale purification of Antibodies or enrichment of Proteins of interest in prepacked spin column, electrophoresis or LC/MS
compliance-icons
Product Identifier
  • Protein G HP SpinTrap
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.