missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein G Mag Sepharose™ Xtra

Product Code. 11555664 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 x 1 mL
5 x 1 mL
Unit Size:
Pack of 2
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11555664 5 x 1 mL Pack of 5
11545664 2 x 1 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11555664 Supplier Cytiva Supplier No. 28967070
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High binding capacity, 27mg human IgG/mL medium, for efficient capture of antibody with high yield and purity

  • Non adherant beads
  • Matrix: paramagnetic, spherical, highly cross-linked agarose particles
  • Ligand: Protein G
  • Color: black
  • Operating temperature: room temperature
  • Can be used without detergents
  • For small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants
  • Available in 2 x 1mL and 5 x 1mL packs

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Protein G Mag Sepharose Xtra magnetic bead
Binding Capacity 27mg human IgG/mL gel
Matrix Paramagnetic, spherical, highly cross-linked agarose particles
Quantity 5 x 1 mL
Content And Storage 4°C to 8°C
For Use With (Application) For rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants
Ligand Protein G
compliance-icons
Product Identifier
  • Protein G Mag Sepharose Xtra
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-66.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.