missing translation for 'onlineSavingsMsg'
Learn More

Protein Gel-Loading Dye for SDS-PAGE, 2X (Dark-Purple Liq./Electrophor.), Fisher BioReagents

Brand:  Fisher Bioreagents BP637-1

Code : 32

Additional Details : Weight : 0.05100kg

 View more versions of this product

Product Code. 10429313

  • 84.83 € / 1mL


Estimated Shipment
15-03-2019
Add to basket

Description

Description

This dye is added to protein samples prior to electrophoresis on SDS–PAGE gels.
Specifications

Specifications

0.15 to 0.35 AU at 252nm, 0.25 to 0.40 AU, 0.32 to 0.47 AU
7647-01-0
Pass Test
Liquid
Poly Tube
Purple
Protein Gel-Loading Dye for SDS-PAGE, 2X
DNase free
Sodium Dodecyl Sulfate (<5%), Glycerol (<12%), Tris (<1.0%), Proprietary Component (<2%), and Water (80%).
Electrophoresis
Protease free
1mL
Videos
SDS Documentation