missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Protein Kinase Inhibitor

Product Code. 15424239
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424239 1 mg 1mg
15414239 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424239 Supplier Thermo Scientific Chemicals Supplier No. J61687.M^
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97161-97-2 | C27H21N3O5, C27H21N3O5 | 467.48 g/mol

Inhibitor of protein kinase A

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97161-97-2
Molecular Formula C27H21N3O5
Synonym K252a

Specifications

Quantity 1 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of protein kinase A
Formula Weight 467.47
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.