missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protein LoBind™ Tubes
GREENER_CHOICE

Product Code. 17134364 Shop All Eppendorf Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
15 mL
2 mL
25 mL
5 mL
50 mL
Packaging:
1 Bag x 500 Tubes
100 Tubes (2 Bags x 50 Tubes)
200 Tubes (2 Bags x 100 Tubes)
200 Tubes (4 Bags x 50 Tubes)
200 Tubes (8 Bags x 25 Tubes)
Closure Type:
Screw Cap
Snap Cap
Max. RCF:
17,000 x g
18,000 x g
19,500 x g
25,000 x g
Unit Size:
Pack of 100
Pack of 200
Pack of 500
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Packaging Closure Type Max. RCF Unit Size
17134364 1.5 mL 1 Bag x 500 Tubes Snap Cap 18,000 x g Pack of 500
17463703 25 mL 200 Tubes (4 Bags x 50 Tubes) Screw Cap 17,000 x g Pack of 200
17124364 0.5 mL 1 Bag x 500 Tubes Snap Cap 18,000 x g Pack of 500
17144364 2 mL 1 Bag x 500 Tubes Snap Cap 18,000 x g Pack of 500
15925180 15 mL 200 Tubes (4 Bags x 50 Tubes) Screw Cap 19,500 x g Pack of 200
15945180 50 mL 200 Tubes (8 Bags x 25 Tubes) Screw Cap 19,500 x g Pack of 200
15178344 5 mL 100 Tubes (2 Bags x 50 Tubes) Snap Cap 25,000 x g Pack of 100
16180371 5 mL 200 Tubes (2 Bags x 100 Tubes) Screw Cap 25,000 x g Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17134364 Supplier Eppendorf™ Supplier No. 0030108442
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.

Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.

  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
TRUSTED_SUSTAINABILITY

Specifications

Closure Material HDPE
Color Colorless
Capacity (Metric) 1.5 mL
Material Polypropylene
No. per Pack 500 Pk.
Packaging 1 Bag x 500 Tubes
Sterility Non-sterile
Autoclavable Autoclavable
Bottom Shape Conical
Closure Type Snap Cap
Product Type Microcentrifuge Tube (PCR Clean, Protein LoBind)
Max. RCF 18,000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.