missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Protein Quant Sample Lysis Kit, 25 mL

Product Code. 11690139 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11690139 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11690139 Supplier Applied Biosystems™ Supplier No. 4448536
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein Quant Sample Lysis Kit, 25 mL

The Protein Quant Sample Lysis Kit contains the reagents required for the preparation of target and control cell lysates for use with TaqMan™ Protein Assay experiments. It contains a one-step lysis solution for preparing crude cell lysates from cultured cells, and the lysates are compatible with cell counting as well as total protein quant measurements such as a microBCA assay. The Protein Quant Sample Lysis Kit contains a buffered nonionic detergent solution for the gentle lysis of adherent and non-adherent cultured cells under non-denaturing conditions
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.