missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Protein Select™ Resin

Product Code. 30284842
Change view
Click to view available options
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
30284842 Each
30284848 Each
30284840 Each
30284828 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30284842 Supplier Cytiva™ Supplier No. 0045CHP01TA

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag.

  • Infrared (IR) turbine rotor reflection ensures accurate and repeatable flow measurements, ideal for precision-critical applications
  • Robust and stable design with four screw gaps allows for secure and easy mounting in various system configurations
  • Constructed from USP Class VI PVDF material, offering excellent chemical resistance and compatibility with aggressive liquids
  • Supports gamma sterilization up to 50kGy, making it suitable for use in sterile environments and medical-grade processes
  • Delivers high-resolution square wave output for reliable signal processing and integration with control systems
  • Tubes can be sterilized up to 150°C (302°F), enabling safe use in high-temperature cleaning and sterilization protocols
  • Designed to reduce downtime and prevent cross-contamination, enhancing operational efficiency and product safety
  • Each sensor is pre-calibrated to achieve accuracy within <1%, minimizing setup time and ensuring consistent performance
TRUSTED_SUSTAINABILITY

Specifications

Type Flowmeter
Accuracy ±1%
Airflow Rate 0.03 to 2 L/min.
Case Material PVDF
Float Material PVDF
Gas Type All
Pressure Rating 362 psig
Includes 7 mm HB Tubeholder System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.