Learn More
Invitrogen™ eBioscience™ Protein Transport Inhibitor Cocktail (500X)
Product Code. 15172069
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 15172069
Brand: Invitrogen™ 00498093
Edge
Suitable for use in assays in which prevention of protein transport to the extracellular space is desired.
  • A cocktail of Brefeldin A and Monensin
  • Suitable for use in assays in which prevention of protein transport to the extracellular space is desired
  • It has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
  • Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus
  • This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting
TRUSTED_SUSTAINABILITY
Specifications
Product Line eBioscience™
Type Protein Transport Inhibitor Cocktail
Quantity 100 μL
compliance-icons compliance-icons
Product Identifier
  • Protein Transport Inhibitor
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol