Learn More
R&D Systems™ Proteome Profiler Human Phospho-Kinase Array Kit
Description
Analyzing the phosphorylation profiles of kinases and their protein substrates is essential for understanding how cells recognize and respond to changes in their environment. The Human Phospho-Kinase Array is a rapid, sensitive, and economical tool to simultaneously detect the relative levels of phosphorylation of 37 kinase phosphorylation sites and 2 related total proteins without performing numerous immunoprecipitations and Western blots. Each capture antibody was carefully selected using cell lysates prepared from cell lines known to express the target protein.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8°C. Do not use past expiration date. |
| Quantity | 1 Kit |
| Target Species | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.