missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Proteome Profiler Human XL Cytokine Array Kit
GREENER_CHOICE

Product Code. 18017641
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18017641 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18017641 Supplier R&D Systems™ Supplier No. ARY022B
  • ONLINE EXCLUSIVE PRICE
    1115.00 €
    1054.20 € / 1 sets
    Save 60.80 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Proteome Profiler Human XL Cytokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.

The Proteome Profiler Human XL Cytokine Array Kit contains 4 membranes - each spotted in duplicate with 105 different cytokine antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Description Cytokine Assay Kits
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit
For Use With (Application) For parallel determination of human cytokines and chemokine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.