missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Proteome Profiler Mouse Apoptosis Array

Product Code. 18067641
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18067641 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18067641 Supplier R&D Systems™ Supplier No. ARY031
  • ONLINE EXCLUSIVE PRICE
    732.00 €
    691.95 € / 1 sets
    Save 40.05 €
    5% Off
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Proteome Profiler Mouse Apoptosis Array requires no specialized equipment and eliminates the need for multiple Western blot experiments.

Contains 8 membranes, each spotted in duplicate with 21 different apoptosis antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2 to 8 °C. Do not use past expiration date
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.