missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Proteome Profiler Mouse XL Cytokine Array
Click to view available options
Quantity:
25 μg
Unit Size:
1 set
Description
The Proteome Profiler Mouse XL Cytokine Array contains 4 membranes-each spotted in duplicate with 111 different cytokine antibodies.
Specifications
Specifications
| For Use With (Application) | For parallel determination of mouse cytokines and chemokines |
| Product Type | Cytokine Array Kits |
| Quantity | 25 μg |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction