missing translation for 'onlineSavingsMsg'
Learn More

Prothoate, TRC

Product Code. 30136791
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136791 250 mg 250mg
30135064 1 g 1g
30003946 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136791 Supplier Toronto Research Chemicals Supplier No. TRCP838810250MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2275-18-5
Molecular Formula C9 H20 N O3 P S2
Molecular Weight (g/mol) 285.36
Synonym Phosphorodithioic acid, O,O-diethyl ester, S-ester with N-isopropyl-2-mercaptoacetamide (6CI,7CI,8CI), Acetamide, N-isopropyl-2-mercapto-, S-ester with O,O-di-Et phosphorodithioate (6CI), American Cyanamid 18,682, FAC, FAC (pesticide), Fac 20, Fak 40, Fostion, O,O-Diethyl S-(N-isopropylcarbamoylmethyl) phosphorodithioate, Prothoat, Prothoate, Trimethoate
IUPAC Name 2-diethoxyphosphinothioylsulfanyl-N-propan-2-ylacetamide
SMILES CCOP(=S)(OCC)SCC(=O)NC(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H20NO3PS2/c1-5-12-14(15,13-6-2)16-7-9(11)10-8(3)4/h8H,5-7H2,1-4H3,(H,10,11)
Formula Weight 285.0622
Chemical Name or Material Prothoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.