missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Lid for Buerkle™ HDPE Universal Storage Containers

Product Code. 10071462 Shop All Bürkle Products
Change view
Click to view available options
Dimensions (L x W x H):
660 x 450 x 220 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10071462 660 x 450 x 220 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10071462 Supplier Buerkle™ Supplier No. 34180001
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lid for Polypropylene Storage and Stacking Container – Polypropylene Storage and Stacking Container with external reinforcement ribs.

  • PS
  • For universal storage containers
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lid
Color White
Material Polystyrene
Closure Type Lid
Description Equipment for universal storage containers
Dimensions (L x W x H) 660 x 450 x 220 mm
For Use With (Equipment) Universal Storage Containers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.