missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PSC 833

Product Code. 18094372
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18094372 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18094372 Supplier R&D Systems™ Supplier No. 4042/1
  • ONLINE EXCLUSIVE PRICE
    396.96 €
    355.95 € / 1mg
    Save 41.01 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of P-gp-mediated MDR

P-glycoprotein (P-gp) modulator; inhibits P-gp-mediated multidrug-resistance (MDR). Reverses resistance to several cytotoxic drugs including mitoxantrone and doxorubicin (resistance factors are 2.0 and 6.5 respectively) in human MDR cancer cell lines. Non-immunosuppressive analog of cyclosporin A (Cat. No. 1101).
TRUSTED_SUSTAINABILITY

Specifications

CAS 121584-18-7
Quantity 1 mg
Product Line Tocris
Molecular Formula C63H111N11O12
Formula Weight 1214.62
Purity >98%
Molecular Weight (g/mol) 1214.65
Chemical Name or Material PSC 833
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.