missing translation for 'onlineSavingsMsg'
Learn More

Pseudopelletierine, Thermo Scientific Chemicals

Product Code. 16524404 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16524404 1 g 1g
16534404 5 g 5g
16544404 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16524404 Supplier Thermo Scientific Acros Supplier No. 463980010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-70-5
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.23
InChI Key RHWSKVCZXBAWLZ-UHFFFAOYNA-N
IUPAC Name 9-methyl-9-azabicyclo[3.3.1]nonan-3-one
SMILES CN1C2CCCC1CC(=O)C2

Specifications

Quantity 1 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 153.22
Chemical Name or Material Pseudopelletierine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.