missing translation for 'onlineSavingsMsg'
Learn More

β-Pseudouridine, MP Biomedicals™

Product Code. 11491212 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491212 50 mg 50mg
11481212 10 mg 10mg
11401222 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491212 Supplier MP Biomedicals Supplier No. 0219917850
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in initiation of translation. β-Pseudouridine, MP Biomedicals is an isomer of the nucleoside uridine, found in all species and in many classes of RNA (except mRNA). Studies suggest that it reduces radiation-induced chromosome aberrations in huma

β-Pseudouridine is formed by enzymes called Ψ synthases, which post-transcriptionally isomerize specific uridine residues in RNA in a process termed pseudouridylation.

Chemical Identifiers

CAS 1445-07-4
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.20
MDL Number MFCD00038458
InChI Key PTJWIQPHWPFNBW-GBNDHIKLSA-N
Synonym pseudouridine, beta-pseudouridine, psi-uridine, 5-beta-d-ribofuranosyl uracil, beta-d-pseudouridine, 5-b-d-ribofuranosyl-uracil, pseudouridine c, 5-ribosyluracil, 1s-1,4-anhydro-1-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl-d-ribitol, 5-2s,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione
PubChem CID 15047
ChEBI CHEBI:17802
IUPAC Name 5-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)C1=CNC(=O)NC1=O

Specifications

Density 1.64g/cm3
Boiling Point 598.4°C (760 mmHg (literature))
Refractive Index 1.62 (Lit.)
Quantity 50 mg
Solubility Information Soluble in water (50mg/mL, clear, colorless solution).
Formula Weight 244.2g/mol
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.