missing translation for 'onlineSavingsMsg'
Learn More

PSF™ Cofra Tullus Safety Shoes

Product Code. 15296448
Change view
Click to view available options
Size:
3
4
5
6
10
11
12
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15296448 12 Each
15285118 5 Each
15216448 10 Each
15225128 6 Each
15255008 4 Each
15255118 3 Each
15276448 11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15296448 Supplier PSF™ Supplier No. TULLUSSIZE12

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color White
For Use With (Application) Canteens, Food and Chemicals Industries, Chemistry, Hospital, Clinic
Liner Material Sany-Dry
Size 12
Sole Features Dual Density
Sole Material Polyurethane
Style Tullus
Resistant To Rips, abrasions, tears and scratches
Waterproof Yes
Product Type Cofra Tullus Safety Shoes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.