missing translation for 'onlineSavingsMsg'
Learn More

PSF™ Contractor Safety Shoes

Product Code. 12887573
Change view
Click to view available options
Size:
6
7
8
9
10
13
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12887573 10 1 pair
12857583 9 1 pair
12847583 8 1 pair
12807553 6 1 pair
12817583 13 1 pair
12837583 7 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12887573 Supplier PSF™ Supplier No. 806SM10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance S1 Rated
Color Black
For Use With (Application) Industry, Maintenance, College, Mechanics
Material Leather
Size 10
Sole Features Dual Density
Sole Material Polyurethane
Style Contractor Safety
Toe Material Steel
Resistant To Heat, oils and general chemicals
Product Type Contractor Safety Shoe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.