Learn More
Hamilton PT1000 Probe Support Stand
Product Code. 31257792
Change view
Click to view available options
Material:
Stainless Steel
Product Code. Material
31257792 Stainless Steel
1 options
Product Code. 31257792 Supplier Hamilton Supplier No. 10176378
  • ONLINE EXCLUSIVE PRICE
    244.00 €
    216.04 € / Pack of 1
    Save 27.96 €
    11% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides a stable and secure platform for the PT1000 temperature probe when used with a magnetic heater stirrer. Ensures reliable positioning during laboratory temperature monitoring.
  • Adjustable PT1000 temperature probe support stand
  • Attaches securely to the MagHeat magnetic stirrer
  • Keeps the temperature probe stable during operation
Specifications
Type Temperature Probe
Warranty None
Material Stainless Steel