missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Tube

Product Code. 16309571 Shop All Bürkle Products
Change view
Click to view available options
Flow Rate:
180 mL/stroke
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
16309571 180 mL/stroke Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16309571 Supplier Buerkle™ Supplier No. 56060400
  • ONLINE EXCLUSIVE PRICE
    674.86 €
    622.35 € / Pack of 1
    Save 52.51 €
    7% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The PTFE Ultrapure Barrel Pump with Discharge Tube is suitable for the transfer of ultra-pure liquids and those that attack conventional materials such as PP or PVC. The stopcock prevents vapours from escaping, and stops dripping after transfer.

  • Barrel pump with rigid discharge tube with stopcock made of PTFE
  • High chemical resistance
  • Also suitable for transferring ultra-pure liquids
  • The stopcock prevents hazardous vapours from escaping, and stops dripping after transfer
  • Immersion tube Ø 32 mm
  • All valves are made of solid PTFE/PVDF
  • Top quality - 3 years' warranty
  • Barrel screw joints for commercially available containers available as accessories
TRUSTED_SUSTAINABILITY

Specifications

Applications For transferring ultra-pure liquids
Description Barrel pump with rigid discharge tube with stopcock made of PTFE
Pump Head Material PTFE
Flow Rate 180 mL/stroke
Depth (Metric) 40 cm
Type Barrel Pump
Certifications/Compliance Material certificate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.