Learn More
Kartell™ Lining for Glass Tapers
Description
- Designed to prevent machined glass joints from binding
- Conforms to DIN 12242
- Suitable for foodstuff
Specifications
Specifications
| Certifications/Compliance | DIN 12242 |
| Material | PTFE |
| Diameter (Metric) Outer | 12 mm |
| For Use With | Glass Joint |
| For Use With (Application) | Prevent glass joints from binding |
| Packaging Quantity | 10/Pk. |
| Tubing Size | 14/23 Cone Size |
| Product Type | Ground Joint Sleeve Lining |