Learn More
Kartell™ Lining for Glass Tapers
Click to view available options
Tubing Size:
14/23 Cone Size
19/26 Cone Size
24/29 Cone Size
29/32 Cone Size
45/40 Cone Size
Unit Size:
Pack of 10
Description
- Designed to prevent machined glass joints from binding
- Conforms to DIN 12242
- Suitable for foodstuff
Specifications
Specifications
| Certifications/Compliance | DIN 12242 |
| Material | PTFE |
| Diameter (Metric) Outer | 45 mm |
| For Use With | Glass Joint |
| For Use With (Application) | Prevent glass joints from binding |
| Packaging Quantity | 10/Pk. |
| Tubing Size | 45/40 Cone Size |