missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Containers

Product Code. 10140342 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
120 mL
240 mL
360 mL
480 mL
1000 mL
2200 mL
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10140342 60 mL Each
10313502 15 mL Each
10323502 30 mL Each
10291952 120 mL Each
10199901 240 mL Each
10724143 360 mL Each
10393882 480 mL Each
10363602 1000 mL Each
10660473 2200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10140342
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Isostatically molded from pure PTFE. Fisherbrand™ PTFE Containers contain thick wall construction.

  • Smooth internal finish
  • Contains crew cap
  • Stackable
  • Autoclavable

Specifications

Capacity (Metric) 60 mL
Color White
Diameter (Metric) Outer Neck 51mm
Diameter (Metric) Inner Neck 47mm
For Use With (Application) Storage and Transport of Hazardous Chemicals
Includes Closure
Height (Metric) With Closure 49mm
Material PTFE
Max. Temperature (Metric) 280°C
Temperature Range (Metric) -200 to +280°C
Thickness (Metric) 9 mm
Transparency No
Weight (Metric) 100 g
Graduations No
Closure Included Yes
Graduated No
Height (Metric) 47 mm
Closure Size 60 mm
Diameter (Metric) 60 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 2 oz.
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Product Type Storage Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.