missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ PTFE Magnetic Stirring Bars Retriever

Product Code. p-8001262 Shop All Brand Products
Click to view available options
Length (Metric):
150 mm
250 mm
350 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10578401

Brand: BRAND™ 137710

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars Retriever has a magnetic core that is fully sealed to prevent leeching.

  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.

  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 8 mm
Length (Metric) 250 mm
Material PTFE
Quantity 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.