missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Platinum Resistance Probe with 1M Cable

Product Code. 11712464 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
100 mm
200 mm
300 mm
400 mm
500 mm
700 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11712464 100 mm Each
11752464 500 mm Each
11762464 700 mm Each
11722464 200 mm Each
11742464 400 mm Each
11732464 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11712464
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Inert to virtually all chemical reagents. Fisherbrand™ PTFE Thermocouple Type K Probe combine the outstanding properties of PTFE with the accuracy and reliability of platinum resistance and thermocouple thermometry.

  • Non-contaminating
  • Impervious to chemical diffusion at high pressure
  • Wide operating temperature range, -180 to +300°C
  • Outstanding electrical insulation properties
  • Individually tested

Specifications

Type Probe, PTFE Platinum Resistance
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Material PTFE with Stainless Steel Core
Sensor Types PRT100 Class A. TCR = 3850 ppm/degree celsius
For Use With (Application) Measuring temperature
For Use With (Equipment) PTFE Probe Holders and Compression Fittings
Response Time 50%, 6 to 14 sec.
Temperature Range (Metric) -180°C to +280°C
Length (Metric) 100 mm
Width (Metric) 7 mm
Shipping Weight (Metric) 60 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.