missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Fortuna™ Optifix™ Basic Dispenser

Product Code. 10364591
Change view
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
100 to 500 mL
2 to 10 mL
20 to 100 mL
40 to 200 mL
6 to 30 mL
60 to 300 mL
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10364591 0.4 to 2 mL Each
10339800 100 to 500 mL Each
10518151 2 to 10 mL Each
10068790 1 to 5 mL Each
10768751 10 to 50 mL Each
10577951 20 to 100 mL Each
10633971 40 to 200 mL Each
10008830 6 to 30 mL Each
10798941 60 to 300 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10364591 Supplier Poulten Graf™ Supplier No. 101 08027
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Poulten Graf™ Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.

For dosing of the most common chemicals, acids and crytallizing liquids (not suitable for solvents). Poulten Graf™ PTFE/PP/Fiberglass/Teflon/Ceramic Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.

  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With solid PTFE encased dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment

Specifications

Product Type Analog Bottletop Dispenser (Basic)
Color Clear
Channels 1
Material PTFE, Polypropylene, Fiberglass, Teflon, Ceramic
Volume (Metric) 0.4 to 2 mL
For Use With (Application) For dispensing aqueous solutions, acids and crystallizing liquids
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.