missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Fortuna™ Optifix™ Basic Dispenser
Description
For dosing of the most common chemicals, acids and crytallizing liquids (not suitable for solvents). Poulten Graf™ PTFE/PP/Fiberglass/Teflon/Ceramic Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
- Manufactured according to DIN EN ISO 9001:2008 using high quality materials
- With solid PTFE encased dispensing piston made of glass
- Cylinder coated with plastic to protect against breakage
- With certificate, individually numbered
- Ergonomic finger protection cap made of PP with volume setting system
- Graduation on the scale is lasered and easy to read
- Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
| Product Type | Analog Bottletop Dispenser (Basic) |
| Color | Clear |
| Channels | 1 |
| Material | PTFE, Polypropylene, Fiberglass, Teflon, Ceramic |
| Volume (Metric) | 20 to 100 mL |
| For Use With (Application) | For dispensing aqueous solutions, acids and crystallizing liquids |
| Quantity | Each |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?