missing translation for 'onlineSavingsMsg'
Learn More

PTFE rod, 16mm (0.63 in.) dia, Thermo Scientific™

Product Code. 15422187
Change view
Click to view available options
Quantity:
30 cm
Packaging:
Tube
Unit Size:
30cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15422187 30 cm Tube 30cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15422187 Supplier Thermo Scientific Alfa Aesar Supplier No. 045173.BV
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9002-84-0
Packaging Tube
Molecular Formula (C2F4)n
MDL Number MFCD00084455
Quantity 30 cm
SMILES FC(F)(-*)C(F)(F)-*
Molecular Weight (g/mol) NaN
Chemical Name or Material PTFE rod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.