Learn More
Thermo Scientific™ I-Chem Septa for 24-414 Open Top Caps
Product Code. 12897030
Change view
Click to view available options
Material:
PTFE-lined Silicone, Silicone
Unit Size:
Pack of 72
Product Code. Material Unit Size
12897030 PTFE-lined Silicone, Silicone Pack of 72
1 options
Product Code. 12897030 Supplier Thermo Scientific™ Supplier No. 2887222
  • ONLINE EXCLUSIVE PRICE
    101.00 €
    91.90 € / Pack of 72
    Save 9.10 €
    9% Off
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Loose septa may be used as replacement parts for VOA vials and bottles with unbonded septa. Fits the standard 24-414 open top cap. Processed to meet EPA Performance Based Specifications for Volatile Organic Analysis.
  • PTFE-lined silicone septum
  • 0.125 in. thick or 0.060in. thin natural PTFE-lined silicone septum, 24mm diameter
  • For use with Thermo Scientific VOA vials and bottles with 24-414 open-top cap
  • Round shape,  Natural color
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE-lined Silicone, Silicone
No. per Pack 72Pack
Packaging 72Pack
Quantity 72Pack
Product Type Septa