missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Test Tube Rack

Product Code. 10659112 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
Tubes (13 mm)
Tubes (19 mm)
Tubes (30 mm)
Tubes (8 mm)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
10659112 Tubes (30 mm) Each
10241852 Tubes (13 mm) Each
10467742 Tubes (8 mm) Each
10241902 Tubes (19 mm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10659112
Estimated Shipment: 26-08-2026
Add to basket
Add to basket

Use from cryogenic to 250°C (480°F). Fisherbrand™ PTFE Test Tube Rack is suitable for ovens and high temperature themostats.

  • Inert to virtually all chemical reagents
  • Suitable for repeatedly sterilizing

Specifications

Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Autoclavable Autoclavable
Color White
Holds Tubes (30 mm)
Material PTFE
Positions 4
For Use With (Equipment) Test Tubes
Quantity 1 Each
Temperature (Metric) Range -200°C to +280°C
Dimensions (L x W) 180 x 60mm
Dimensions (L x W x H) 180 x 60 x 80mm
Length (Metric) 180 mm
Height (Metric) Interior 70 mm
Height (Metric) Shelf 5 mm
Depth (Metric) 60 mm
Diameter (Metric) Opening 30 mm
Weight (Metric) 256 g
Product Type Test Tube Rack
Height (Metric) 80 mm
Width (Metric) 60 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.