missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Hose

Product Code. 11911591 Shop All Bürkle Products
Change view
Click to view available options
Flow Rate:
270 mL/stroke
400 mL/stroke
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
11911591 400 mL/stroke Each
11901591 270 mL/stroke Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11911591 Supplier Buerkle™ Supplier No. 56061001
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Provides high chemical resistance and safe transfer, even with liquids which attack conventional materials such as PP or PVC.

  • Medium-contacting parts manufactured from PTFE
  • Suitable for transferring ultra-pure liquids
  • Stopcock prevents hazardous vapours from escaping and stops dripping after transfer
  • Discharge hose is 1.2m in length
  • Immersion tube with a diameter of 32mm
  • All valves are made of solid PTFE/PVDF
  • Top quality, 3 years’ warranty
TRUSTED_SUSTAINABILITY

Specifications

Description Immersion depth of 95 cm
Includes Discharge Tube
No. per Pack 1 Ea.
Material PTFE
Flow Rate 400 mL/stroke
Type Barrel Pump
Applications Liquids which attack conventional materials such as Polypropylene or PVC can now be safely transferred (except for elemental Fluorine), transferring ultra-pure liquids
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.