missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Vial

Product Code. 10762793 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
25 mL
50 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10762793 25 mL Each
10650283 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10762793
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Thick wall construction. Fisherbrand™ PTFE Vials are ideal for storing aggressive or expensive materials.

  • Suitable for small scale reactions at low pressure
  • Tapered inner for easy removal of contents

Specifications

Diameter (Metric) Inner 25 mm
Color White
Diameter (Metric) Outer 35 mm
Material PTFE
Volume (Metric) 25 mL
For Use With (Application) Storage of hazardous chemicals
Graduated No
Temperature Range (Metric) -200°C to +280°C
Closure Type Screw Cap
Quantity 1 Ea.
Closure Size 45 mm
Autoclavable Autoclavable
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Height (Metric) 64 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.