Learn More
VITLAB™ PTFE Watch Glass
Product Code. 10080731
Change view
Click to view available options
Diameter (Metric):
50 mm
Unit Size:
Each
Product Code. Diameter (Metric) Unit Size
10080731 50 mm Each
1 options
Product Code. 10080731 Supplier VITLAB™ Supplier No. 113197
  • ONLINE EXCLUSIVE PRICE
    14.00 €
    11.47 € / Pack of 1
    Save 2.53 €
    18% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Autoclavable Autoclavable
Material PTFE
Shape Round
Type Watch Glass
For Use With (Application) Used as Beaker Covers
Diameter (Metric) 50 mm