missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ PTFE Watch Glass

Use a watch glass to evaporate liquids, hold solids when weighing a sample, and in situations where cross-contamination could occur. Kartell™ PTFE Watch Glass has good heat and chemical resistance.

Brand:  Kartell™ 0064400

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10293991

  • 30.30 € / Pack of 2
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

  • Superior chemical, mechanical, and heat resistance
  • Can be used safely for continuous service at temperatures up to 250°C
  • Suitable for foodstuff
Specifications

Specifications

Round
Round
76.32 mm
PTFE
Watch Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers