missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ PTFE Watch Glass
Use a watch glass to evaporate liquids, hold solids when weighing a sample, and in situations where cross-contamination could occur. Kartell™ PTFE Watch Glass has good heat and chemical resistance.
19.85 € - 77.15 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10224141
|
Kartell™
0064700 |
153.92 mm |
77.15 €
Pack of 2 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
10214141
|
Kartell™
0064600 |
126.36 mm |
60.55 €
Pack of 2 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
10707721
|
Kartell™
0064300 |
52.76 mm |
19.85 €
Pack of 2 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
10293991
|
Kartell™
0064400 |
76.32 mm |
30.30 €
Pack of 2 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
10254041
|
Kartell™
0064500 |
99.39 mm |
45.90 €
Pack of 2 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
Description
- Superior chemical, mechanical, and heat resistance
- Can be used safely for continuous service at temperatures up to 250°C
- Suitable for foodstuff
Specifications
PTFE |