Learn More
Fisherbrand™ PTFE Wing Stir Bars
Provides stable stirring action. The PTFE wing stir bars are effective in small tapered and round bottom test, centrifuge, and sample tubes.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 16265781
  • 37.00 € / Pack of 10
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • For effective stirring in small tapered and round bottom test, centrifuge, and sample tubes
  • Autoclavable
Specifications
Specifications
13 mm
20mm
14-15mm diameter test or centrifuge tubes; Test, Centrifuge & Sample Tubes
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
White
Yes
Wing