missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ PU First Grey Gloves

Product Code. 12655293
Click to view available options
Size:
6
10
11
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12655293

Brand: Honeywell Safety Products™ 210025011

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Grey polyamide knitted gloves with polyurethane-coated palm and fingertips provide dexterity, a snug fit, and good grip for precision work. Honeywell Safety Products™ PU First Grey Gloves are knitted for breathability and comfort and offer excellent abrasion and tear resistance.

  • Grey, non-pilling polyamide knitted glove suitable for precision work.
  • Suitable for handling fine products or dirty objects in dry, slightly greasy or dirty environments.
  • Excellent abrasion and tear resistance.
  • Silicone-free and suitable for painting environments.
  • Grey polyurethane coating on palm and fingertips for good grip.
  • Tight knit provides comfort, support, dexterity, touch and limits dirt penetration.
  • Ventilated back optimizes breathability.
  • Elastic wrist.
  • Grey color enhances soil proof qualities, ensuring longer life.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Minor risks, EN 420-2003 Basic standard, EN 388-2003 Mechanical risk, EC Category PPE 2, EC Certificate Number 0075/797/162/11/06/0281, EC Attestation Number 0075/797/162/11/06/0281
Classification Dexterity: 5
Coated Area Palm, Fingertips
Construction Seamless Knit
Description Gauge 15
Coating Material Polyurethane
Color Gray
Cut Resistance Rating 1
Material Polyamide
Product Type Gloves
Size 11
For Use With (Application) Service, Telecom, Transportation and Utility industries
Includes Elasticized wrist
Puncture Rating 1
Quantity 10Pair/PK
Tear Rating 3
Length (Metric) 25 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.