Learn More
Chemsplash PU Grip Anti-Slip Sole OverShoe
Product Code. 30058269
Change view
Click to view available options
Size:
Universal
Unit Size:
200 pairs
Product Code. Size Unit Size
30058269 Universal 200 pairs
1 options
Product Code. 30058269 Supplier Chemsplash Supplier No. 2782.400.007
  • ONLINE EXCLUSIVE PRICE
    347.19 €
    322.55 € / 200 pairs
    Save 24.64 €
    7% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The Grip Overshoe with Anti-Slip sole is made from Chemsplash 55 gsm micro laminate fabric.
Chemsplash Grip Overshoe
  • 55GSM Type PB 6B Material
  • 70gsm Grey PU Anti-Slip Sole
  • Elasticated Top and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Grip Overshoe with Anti-Slip sole is made from Chemsplash 55gsm Micro Laminate fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This overshoe is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforming to ISO 14644-1:2015 Cleanroom Class 5-9, it features a 70gsm PU Anti-Slip Sole that is also anti-static. Chemsplash accessories are designed to protect the wearer from the environment or vice versa, providing maximum comfort and wearability.
Specifications
Size Universal
Material Microporous Composite Fabric, Proprietary Laminated Non-woven Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-slip
Resistance Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)