missing translation for 'onlineSavingsMsg'
Learn More

Ansell PU800 Medium Duty Cut-Resistant Polyurethane-Coated Gloves

Product Code. 16835290
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16835290

Brand: Ansell B092090

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Polyurethane-coated palm provides durability, and enhances grip capabilities for secure handling in light oil environments.

  • Polyurethane-coated palm enhances grip capabilities in light oil environments
  • Cut resistant technology provides ANSI Level A2 protection
  • Seamless polyethylene/polyester 13-gauge liner
  • High degree of flexibility and dexterity
  • Latex knit wrist ensures a snug fit
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
Material Polyethylene
Size 9
For Use With (Application) Automotive Assembly, Metal Fabrication, Machinery, and Equipment Operation
Glove Cuff Style Knit
Series PU800
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.