missing translation for 'onlineSavingsMsg'
Learn More

Pumice Stone, Technical, Granular, 7-9mm, Fisher Chemical

Product Code. 10676842 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10676842 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10676842 Supplier Fisher Chemical Supplier No. P/7800/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Physical Form Solid
Packaging Plastic Powder Jar
Color Gray
Quantity 500 g
CAS 1332-09-8
Molecular Formula Al2O5Si
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370732 MFCD07370733
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILES O=[Si]=O.O=[Al]O[Al]=O
IUPAC Name oxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular Weight (g/mol) 162.04
PubChem CID 76973482
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.