missing translation for 'onlineSavingsMsg'
Learn More

Pure Distilled Water, Waternation

Product Code. 30130304 Shop All Waternation Medical Division Products
Change view
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
6 x 1000 mL
Unit Size:
1 L
5 L
6 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30130304 10 x 500 mL 5 L
30130307 10 x 100 mL 1 L
30130308 6 x 1000 mL 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30130304 Supplier Waternation Supplier No. MLG3500/10
  • ONLINE EXCLUSIVE PRICE
    82.16 €
    74.61 € / 5 L
    Save 7.55 €
    9% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile Pure Distilled Water which complies with the requirements of ASTM D1193-06.

  • Ready to use
  • No chemical additives
  • Batch traceability
  • Certificate of conformity and analysis
  • Reduce expensive test fails due to impurities
  • High performance
  • High accuracy
  • Suppress water-forming side reactions
  • Provide consistent performance across many sample types

The MLG range, a comprehensive suite of five high grade pure waters, in assorted sizes, ranging from simple deionised to molecular biology grades, catering to the majority of pure water lab uses.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH 7
Conductivity 0.1 to 1 μS/cm
Grade Pure Distilled Water
Packaging PET Plastic Bottle
Certificate of Analysis Yes
Quantity 10 x 500 mL
For Use With (Application) Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.