missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Pure-Fit SIB Polypropylene

Product Code. p-200080384
Click to view available options
Diameter (Metric) Inside:
1/8 to 3/8 in.
3/8 to 3/4 in.
Quantity:
25/Pk.
250/Pk.
Unit Size:
Pack of 25
Pack of 250
This item is not returnable. View return policy

Product Code. 30040250

Brand: Saint Gobain Life Sciences PFTC375

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pure-Fit TC's innovative design allows for complete flow stoppage, and the clamp can be installed over the tube in the normal fashion, or put together after the tube is already part of a completed assembly.

Pure-Fit™ TC tube clamps represent a new generation of clamping technology for the pharmaceutical industry. They feature an outer body with no sharp edges, eliminating the potential for snags, scratches, or punctures; a press-down locking system that is fully encapsulated to guard against untimely or unwanted release; a side release mechanism that facilitates secure, single-handed installation and operation; and an integral bore hole for absolute lock-out capabilities.

  • Provides complete fluid stoppage
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Single-hand installation capability for ease of operation
  • Unique top locking mechanism and side release prevent unwanted openings
  • Available in PVDF or polypropylene
  • Fully autoclavable and sterilizable
  • Custom colors available

Applications

  • Pharmaceutical Processes
  • Peristaltic Pump Sets
  • Drug Delivery and Discovery
  • Medical Systems
  • Laboratory Functions
  • Assemblies and Tubing Sets
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Quantity 25/Pk.
Type Tubing Clamp
Material Polypropylene
Color Clear
Jaw Type Standard Taper
Diameter (Metric) Inside 1/8 to 3/8 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.