missing translation for 'onlineSavingsMsg'
Learn More

DURAN™ PURE Silicone PTFE-coated Seal
BIOPHARMA

Product Code. 16424954 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 25
GL 45
GLS 80
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
16424954 GL 25 Pack of 10
16434954 GL 45 Pack of 10
16444954 GLS 80 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16424954 Supplier DURAN™ Supplier No. 292481456
  • ONLINE EXCLUSIVE PRICE
    39.20 €
    37.02 € / Pack of 10
    Save 2.18 €
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-quality silicone seals developed for the needs of the pharmaceutical and biotech industry

  • High-quality silicone seals developed for the needs of the pharmaceutical and biotech industry
  • High-quality, PTFE coated silicone seal for use with DURAN™ PURE Premium Screw Cap, GL 25
  • Packaged in clean room
  • Single batch delivery
  • Certificate of Analysis (CoA)
  • Visual and dimensional inspection
  • Change control management and information of customer in case of changes
  • USP <88> Class VI Pharmaceutical grade materials

Specifications

Color Light Ivory, Gray
Description PTFE-coated silicone seal
Material Platinum-cured Silicone, PTFE
Thickness (Metric) 3 mm
Thread Size (Metric) GL 25
For Use With (Equipment) GL 25 cap
Diameter (Metric) 23.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.