Learn More
Veolia Water Systems Purelab Flex 3
Product Code. 13135970
Change view
Click to view available options
PURELAB flex 3; DNase <20 pg/ml
Product Code.
13135970 PURELAB flex 3; DNase <20 pg/ml
1 options
Product Code. 13135970 Supplier Veolia Water Systems Supplier No. PF3XXXXM1
  • ONLINE EXCLUSIVE PRICE
    15041.17 €
    9402.87 € / Pack of 1
    Save 5638.30 €
    37% Off
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Rich, reliable, robust, and easy to use ultimate system providing tap to ultrapure water in one single unit.
  • The PURELAB flex 3 is the ultimate system providing tap to ultrapure water in one single unit
  • Is uniquely designed around your needs
  • Combines ease of use with the accurate delivery of pure water
  • The flexible modular system can be configured to deliver the quality of water you need for your applications
  • Daily volume requirements Type I ultrapure water <10L Type III RO water <10L 230V 50/60Hz 236mm x 374mm x 1020mm (w x d x h) Elga
  • Dispense flowrate    Up to 2.0 l/min    Up to 2.0 l/min; Inorganics (resistivity at 25°C); Organics (TOC)    <5 ppb; Bacteria    <0.1 CFU/ml ; Bacterial endotoxin    <0.001 EU/ml; pH    Effectively neutral; DNase    <20 pg/ml; RNase    <0.002 ng/ml