missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Expi Endotoxin-Free Maxi Plasmid Purification Kit

Product Code. p-7180375
Click to view available options
Quantity:
10 Preps
25 Preps
4 Preps
Unit Size:
25 preps
Pack of 10
Pack of 4
This item is not returnable. View return policy

Product Code. 15756003

Brand: Invitrogen™ A33073

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange membrane.

The PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange membrane. In as little as 90 minutes, purify up to 1.5 mg of high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.

Advantages of the PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit include:
• Simple and fast protocol—purification typically takes 90 minutes
• High yield—isolate up to 1.5 mg of high quality plasmid DNA
• Endotoxin-free plasmid—less than 0.1 EU/μg, ideal for sensitive applications
• High purity—ultralow levels of protein, RNA, and genomic DNA contamination
• Compatibility—suitable for use with ExpiCHO and Expi293 expression systems
• Endotoxin-free components— elution buffer, water, and plasmid storage tubes that are free of endotoxin

Simple and fast protocol
Unlike other anion-exchange-based endotoxin-free plasmid purification systems, the PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit utilizes centrifugation- or vacuum-assisted filtration for lysate clarification and subsequent plasmid DNA purification. There is no waiting for gravity-flow columns and the entire purification typically takes 90 minutes (including the final precipitation step).

Endotoxin-free plasmid
Purify endotoxin-free (less than 0.1 EU/μg) plasmid that is ideal for sensitive applications such as transfection of primary cells, genome engineering, and research on gene therapy or plasmid vaccines in vivo.

Compatible with ExpiCHO and Expi293 expression systems
Plasmid purified using the PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit is fully compatible with the Gibco ExpiCHO and Expi293 transient mammalian expression systems.

Superior quality with endotoxin-free components
Rest assured that the kit is of high quality and has additional endotoxin-free elution buffer, water, and plasmid storage tubes for added peace of mind.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Resuspension buffer
• Lysis indicator
• RNase A
• Lysis buffer
• Precipitation buffer
• Endotoxin removal buffer
• Wash buffer
• Elution buffer
• TE buffer
• Expi Maxi filtration column
• Expi Maxi DNA binding column
• 50 mL conical tubes

Endotoxin-free components: elution buffer, water, plasmid storage tubes

Format Spin Column
Isolation Technology Anion Exchange Resin
Scale Maxi
Sample Type Bacteria
Expression System Mammalian
For Use With (Application) Cloning, PCR
High-throughput Compatibility Manual Protocols
No. of Reactions 4 Preps
Prep Scale > 1 mg (Large-Scale) Plasmid DNA, > 200 μg (Large-Scale) Plasmid DNA
Product Line PureLink
Product Type Plasmid Purification Kit
Quantity 4 Preps
Shipping Condition Room Temperature
Target Plasmid DNA
Test Time 90 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.