missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Expi Endotoxin-Free Plasmid Purification Kits

Product Code. 15654154
Change view
Click to view available options
Product Type:
Plasmid Purification Kit
Quantity:
2 Preps
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
15654154 Plasmid Purification Kit 2 Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15654154 Supplier Invitrogen™ Supplier No. A31233
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Resuspension Buffer
  • Lysis Indicator
  • RNase A
  • Lysis Buffer
  • Precipitation Buffer
  • Endotoxin Removal Buffer
  • Wash Buffer
  • Elution Buffer
  • TE Buffer
  • Expi Giga Filtration Column
  • Expi Giga DNA Binding Column
  • Endotoxin-free components: elution buffer, water, plasmid storage tubes

The PureLink™ Expi Endotoxin-Free Plasmid Purification Kits enable quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin.

The PureLink™ Expi Endotoxin-Free Plasmid Purification Kits enable quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin.

The PureLink Expi Endotoxin-Free Plasmid Purification Kits enable quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange membrane. In as little as 2 hours, purify up to 15 mg of high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.

Advantages of the PureLink Expi Endotoxin-Free Plasmid Purification Kits include:

  • Simple and fast protocol—purification typically takes 2 hours
  • High yield—isolate up to 15 mg of high quality plasmid DNA
  • Endotoxin-free plasmid—less than 0.1 EU/μg, ideal for sensitive applications
  • High purity—ultralow levels of protein, RNA, and genomic DNA contamination
  • Compatibility—suitable for use with ExpiCHO and Expi293 expression systems
  • Endotoxin-free components— elution buffer, water, and plasmid storage tubes that are free of endotoxin

Simple and fast protocol

Unlike other anion-exchange-based endotoxin-free plasmid purification systems, the PureLink Expi Endotoxin-Free Plasmid Purification Kits utilize centrifugation- or vacuum-assisted filtration for lysate clarification and subsequent plasmid DNA purification. There is no waiting for gravity-flow columns and the entire purification typically takes 2 hours (including the final precipitation step).

Endotoxin-free plasmid

Purify endotoxin-free (less than 0.1 EU/μg) plasmid that is ideal for sensitive applications such as transfection of primary cells, genome engineering, and research on gene therapy or plasmid vaccines in vivo.

Compatible with ExpiCHO and Expi293 expression systems

Plasmid purified using the PureLink Expi Endotoxin-Free Plasmid Purification Kits are fully compatible with the Gibco ExpiCHO and Expi293 transient mammalian expression systems.

Superior quality with endotoxin-free components

Rest assured that the kits are of high quality and have additional endotoxin-free elution buffer, water, and plasmid storage tubes for added peace of mind.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• Resuspension Buffer
• Lysis Indicator
• RNase A
• Lysis Buffer
• Precipitation Buffer
• Endotoxin Removal Buffer
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi Giga Filtration Column
• Expi Giga DNA Binding Column

Endotoxin-free components: elution buffer, water, plasmid storage tubes

Format Spin Column
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
Expression System Mammalian
For Use With (Application) Cloning, PCR
High-throughput Compatibility Manual Protocols
No. of Reactions 2 Preps
Prep Scale Large Volume Microbial Samples (> 10 mL)
Product Line PureLink
Product Type Plasmid Purification Kit
Quantity 2 Preps
Shipping Condition Room Temperature
Target Endotoxin-free Plasmid
Test Time 2 hr.
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.