missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Fast Low-Endotoxin Maxi Plasmid Purification Kit

Product Code. 15801782
Click to view available options
No. of Reactions:
10 Preps
20 Preps
Unit Size:
Pack of 10
Pack of 20
This item is not returnable. View return policy

Product Code. 15801782

Brand: Invitrogen™ A35895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PureLink Fast Low-Endotoxin Maxi Plasmid Purification Kit uses next-generation columns with advanced silica membranes that enable isolation of transfection grade (<1 EU/μg) plasmid DNA in about 30 minutes.

The simple precipitation-free protocol yields up to 1.5 mg plasmid, suitable for standard transfections and all molecular biology applications such as cloning and sequencing.

  • Ultra-fast protocol—innovative silica-based columns enable purification of transfection grade plasmid DNA in under 30 minutes
  • Low endotoxin plasmid—the resulting plasmid contains less than 1 EU/μg endotoxins
  • High yields—isolate up to 1.5 mg of high-quality plasmid DNA
  • Colored buffers—the kit features colored buffers that permit error-free visualization of complete bacterial cell lysis and subsequent neutralization
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Resuspension buffer, store at 4°C upon reciept
• Lysis buffer, store at room temperature
• Precipitation buffer, store at room temperature
• Binding buffer, store at room temperature
• Wash buffer 1, store at room temperature
• Wash buffer 2 (concentrate), store at room temperature
• Elution buffer, store at room temperature
• Column assemblies, store at room temperature
• Syringe filters, store at room temperature
• Syringe plungers, store at room temperature
• Collection tubes, store at room temperature

Format Kit
Isolation Technology Silica Column
Scale Maxi
For Use With (Application) Cloning, PCR
No. of Reactions 10 Preps
Prep Scale Maxi input culture of 100-500 mL to obtain a yield of up to 850 μg
Product Line PureLink
Product Type Plasmid Purification Kit
Quantity 10 Preps
Shipping Condition Room Temperature
Test Time 30 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.